missing translation for 'onlineSavingsMsg'
Learn More

1-Octanethiol, 98%

Product Code. 11397176
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397176 100 mL 100mL
11307186 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11397176 Supplier Thermo Scientific Alfa Aesar Supplier No. L07195.AE
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 111-88-6 | C8H18S | 146.29 g/mol

1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-88-6
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00004912
InChI Key KZCOBXFFBQJQHH-UHFFFAOYSA-N
Synonym 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol
PubChem CID 8144
IUPAC Name octane-1-thiol
SMILES CCCCCCCCS

Specifications

Melting Point -49°C
Density 0.843
Boiling Point 197°C to 199°C
Flash Point 68°C (154°F)
Refractive Index 1.455
Quantity 100 mL
UN Number UN3082
Beilstein 1733101
Solubility Information Not miscible in water.
Formula Weight 146.29
Percent Purity 98%
Chemical Name or Material 1-Octanethiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Octanethiol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: 1-Octanethiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.