missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, ≥99%, ACS Spectrophotometric Grade, Honeywell Riedel-de Haën™

Product Code. 15675090
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass bottle
Unit Size:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15675090 2 L Glass bottle 2 L
15665090 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15675090 Supplier Honeywell Riedel-de Haën Supplier No. 3605622L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Spectrophotometric Grade,≥99%

  • ACS grade quality reagents and solvents tested according to the specific use for spectrophotometric applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -15°C
Density 0.827g/mL at 25°C
Boiling Point 194°C
Flash Point 80°C (176°F)
Packaging Glass bottle
Linear Formula CH3(CH2)7OH
Refractive Index 1.429
Quantity 2 L
Beilstein 1697461
Vapor Density 4.5 (vs air)
Formula Weight 130.23g/mol
Percent Purity ≥99%
Grade ACS
Chemical Name or Material 1-Octanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.