missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, CHROMASOLV™, for HPLC, ≥99%, Solstice

Product Code. 15622960
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15622960 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15622960 Supplier Honeywell-Chromasolv Supplier No. 2932451L
  • ONLINE EXCLUSIVE PRICE
    108.00 €
    88.73 € / 1 L
    Save 19.27 €
    17% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥99%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

CAS 111-87-5
Absorbance max. 1.0 at 215 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.05 at 250 nm, max. 0.20 at 235 nm, max. 0.50 at 225 nm
CAS Max % 100.0000%
Melting Point -15°C (literature)
Density 0.827 g/mL at 25 °C(lit.)
Boiling Point 194°C
Flash Point 80°C
Packaging Glass Bottle
Molecular Formula C8H18O
Linear Formula CH3(CH2)7OH
MDL Number MFCD00002988
Quantity 1 L
Beilstein 1697461
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILES CCCCCCCCO
IUPAC Name octan-1-ol
Molecular Weight (g/mol) 130.23
PubChem CID 957
ChEBI CHEBI:16188
Formula Weight 130.23
Percent Purity >99%
Grade HPLC
Chemical Name or Material 1-Octanol, CHROMASOLV™, for HPLC, ≥99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.