missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, ReagentPlus™, 99%, Solstice

Product Code. 15611040
Change view
Click to view available options
Quantity:
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
2.5 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15611040 2.5 L Glass Bottle 2.5 L
15621040 20 L Stainless Steel Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15611040 Supplier Fluka Supplier No. 1126152.5L
  • ONLINE EXCLUSIVE PRICE
    160.00 €
    130.82 € / 2.5 L
    Save 29.18 €
    18% Off
 Restricted Item
Estimated Shipment: 01-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -15°C
Color Colorless
Density 0.827g/mL at 25°C
Boiling Point 196°C
Flash Point 80°C (176°F)
Packaging Glass Bottle
Linear Formula CH3(CH2)7OH
Refractive Index n20/D 1.429(lit.)
Quantity 2.5 L
Beilstein 1697461
Vapor Density 4.5 (vs air)
Formula Weight 130.23g/mol
Percent Purity 99%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material 1-Octanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Octan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.