missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, TRC

Product Code. 30412705
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412705 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412705 Supplier Toronto Research Chemicals Supplier No. TRCO23790010ML
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Octanol
CAS 111-87-5
Molecular Formula C8 H18 O
Quantity 10 mL
Synonym 1-Octanol, Octyl alcohol (8CI), 1-Hydroxyoctane, Alfol 8, CO 898, CO 898 (solvent), Caprylic alcohol, Caprylyl alcohol, Heptyl carbinol, Kalcohl 0898, Kalcol 0898, Lorol C 8-98, NSC 9823, Octanol, Octilin, Sipol L8, n-Octan-1-ol, n-Octanol, n-Octyl alcohol
InChI Formula InChI=1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3
SMILES CCCCCCCCO
IUPAC Name octan-1-ol
Molecular Weight (g/mol) 130.23
Formula Weight 130.1358
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.