missing translation for 'onlineSavingsMsg'
Learn More

1-Octyl isothiocyanate, 97%

Product Code. 11431848
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431848 25 g 25g
11421848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431848 Supplier Thermo Scientific Alfa Aesar Supplier No. A14501.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4430-45-9 | C9H17NS | 171.30 g/mol

1-Octyl isothiocyanate can be used to produce 1-(6-methyl-quinolin-2-yl)-3-octyl-thiourea. It is used as intermediates in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octyl isothiocyanate can be used to produce 1-(6-methyl-quinolin-2-yl)-3-octyl-thiourea. It is used as intermediates in pharmaceutical industry.

Solubility
It hydrolyzes in water.

Notes
It is moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-45-9
Molecular Formula C9H17NS
Molecular Weight (g/mol) 171.30
MDL Number MFCD00041139
InChI Key YEZHGQZHWKJPCM-UHFFFAOYSA-N
Synonym octyl isothiocyanate, n-octyl isothiocyanate, octylisothiocyanate, 1-octyl isothiocyanate, octanisothiocyanate, 1-isothiocyanatooctan, octane, 1-isothiocyanato, isothiocyanic acid, octyl ester, n-octylisothiocyanate, acmc-1ajcy
PubChem CID 78161
IUPAC Name 1-isothiocyanatooctane
SMILES CCCCCCCCN=C=S

Specifications

Density 0.88
Boiling Point 124°C to 126°C (7 mmHg)
Flash Point 69°C (156°F)
Odor Pungent
Refractive Index 1.49
Quantity 25 g
UN Number UN2922
Beilstein 1760783
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 171.31
Percent Purity 97%
Chemical Name or Material 1-Octyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.