missing translation for 'onlineSavingsMsg'
Learn More

1-Octylamine, 99%

Product Code. 11421237
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421237 100 g 100g
11431237 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11421237 Supplier Thermo Scientific Alfa Aesar Supplier No. B24193.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 111-86-4 | C8H19N | 129.247 g/mol

1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.

Solubility
Miscible with ethanol, benzene and ethyl acetate. Slightly miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-86-4
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008247
InChI Key IOQPZZOEVPZRBK-UHFFFAOYSA-N
Synonym octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d
PubChem CID 8143
ChEBI CHEBI:7728
IUPAC Name octan-1-amine
SMILES CCCCCCCCN

Specifications

Melting Point -1°C
Density 0.78
Boiling Point 178°C to 179°C
Flash Point 60°C (144°F)
Refractive Index 1.429
Quantity 100 g
UN Number UN2735
Beilstein 1679227
Sensitivity Air Sensitive
Solubility Information Miscible with ethanol,benzene and ethyl acetate. Slightly miscible with water.
Formula Weight 129.25
Percent Purity 99%
Chemical Name or Material 1-Octylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Octylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1-Octanamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.