missing translation for 'onlineSavingsMsg'
Learn More

1-Octyne, 98%

Product Code. 11419663
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419663 100 g 100g
11464150 10 g 10g
11409663 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419663 Supplier Thermo Scientific Alfa Aesar Supplier No. A12440.22
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-05-0 | C8H14 | 110.20 g/mol

1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.

Solubility
Miscible with alcohol, ether and other organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents. Keep away from heat, sparks, sources of ignition and flame.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-05-0
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00009546
InChI Key UMIPWJGWASORKV-UHFFFAOYSA-N
Synonym 1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
PubChem CID 12370
IUPAC Name oct-1-yne
SMILES CCCCCCC#C

Specifications

Melting Point °C to 60°C
Density 0.746
Boiling Point 123°C to 126°C
Flash Point 17°C (62°F)
Assay Percent Range 98%
Linear Formula CH3(CH2)5C≡CH
Refractive Index 1.416
Quantity 100 g
UN Number UN3295
Beilstein 1734494
Solubility Information Miscible with alcohol,ether and other organic solvents. Immiscible with water.
Formula Weight 110.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Octyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.