missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 1-Oleoyl Lysophosphatidic Acid (Sodium Salt)

Product Code. 17120112
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17120112 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17120112 Supplier STEMCELL Technologies Supplier No. 72694
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lysophosphatidic acid (LPA)1 and LPA2 agonist

  • 1-Oleoyl Lysophosphatidic Acid is a species of lysophosphatidic acid (LPA) containing oleic acid at the sn-1 position
  • LPA mediates a variety of biological responses including cell proliferation, smooth muscle contraction, platelet aggregation, neurite retraction, and cell motility (Moolenaar)
  • Stimulates neuronal differentiation in cultured mouse or rat neural progenitor cells (Cui and Qiao; Fukushima et al.; Spohr et al.)
  • Inhibits human embryonic stem (ES) cell-derived neural stem cells (NSCs) from forming neurospheres and differentiating into neurons in vitro (Dottori et al.)
  • Stimulates differentiation of human adipose tissue-derived mesenchymal stem cells to myofibroblast-like cells in vitro (Jeon et al.)

Specifications

CAS 22556-62-3
Synonym Oleoyl-sn-3-glycerophosphate
Quantity 5 mg
Molecular Formula C21H40O7P · Na
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.