missing translation for 'onlineSavingsMsg'
Learn More

1-Oxyl-2,2,5,5-tetramethylpyrroline-3-carboxylate N-Hydroxysuccinimide Ester, TRC

Product Code. 30417260
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30417260 25 mg
30416541 100 mg
30414369 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417260 Supplier Toronto Research Chemicals Supplier No. TRCO87400025MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37558-29-5
Molecular Formula C13 H17 N2 O5
Molecular Weight (g/mol) 281.28
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 1-λ^{1}-oxidanyl-2,2,5,5-tetramethylpyrrole-3-carboxylate
SMILES CC1(C)C=C(C(=O)ON2C(=O)CCC2=O)C(C)(C)N1[O]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C13H17N2O5/c1-12(2)7-8(13(3,4)15(12)19)11(18)20-14-9(16)5-6-10(14)17/h7H,5-6H2,1-4H3
Formula Weight 281.1137
Chemical Name or Material 1-Oxyl-2,2,5,5-tetramethylpyrroline-3-carboxylate N-Hydroxys
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.