missing translation for 'onlineSavingsMsg'
Learn More

1-(p-Methoxyphenyl)-2-propanol, TRC

Product Code. 30479650
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479650 2.5 g 2.5g
30472541 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30479650 Supplier Toronto Research Chemicals Supplier No. TRCM2262352.5G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30314-64-8
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
Synonym 1-(4-Methoxyphenyl)-2-propanol, 2-Hydroxy-1-(4-methoxyphenyl)propane, 2'-Hydroxy-p-propylanisole, p-Methoxy-alpha-methylphenethyl Alcohol, 4-Methoxy-alpha-methylbenzeneethanol
IUPAC Name 1-(4-methoxyphenyl)propan-2-ol
SMILES COc1ccc(CC(C)O)cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C10H14O2/c1-8(11)7-9-3-5-10(12-2)6-4-9/h3-6,8,11H,7H2,1-2H3
Formula Weight 166.0994
Chemical Name or Material 1-(p-Methoxyphenyl)-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.