missing translation for 'onlineSavingsMsg'
Learn More

1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine, TRC

Product Code. 30421463
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421463 50 mg
30414285 100 mg
30420466 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421463 Supplier Toronto Research Chemicals Supplier No. TRCP16100050MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26853-31-6
Molecular Formula C42H82NO8P
Molecular Weight (g/mol) 760.08
Synonym 3,5,8-Trioxa-4-phosphahexacos-17-en-1-aminium, 4-hydroxy-N,N,N-trimethyl-9-oxo-7-[[(1-oxohexadecyl)oxy]methyl]-, inner salt, 4-oxide, (7R,17Z)-, 3,5,8-Trioxa-4-phosphahexacos-17-en-1-aminium, 4-hydroxy-N,N,N-trimethyl-9-oxo-7-[[(1-oxohexadecyl)oxy]methyl]-, hydroxide, inner salt, 4-oxide, [R-(Z)]-, Choline, hydroxide, dihydrogen phosphate, inner salt, ester with 1-palmito-2-olein, L- (8CI), 1-Hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phosphocholine, 1-Palmitoyl-2-oleoyl-3-sn-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-3-sn-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-L-α-lecithin, 1-Palmitoyl-2-oleoyl-L-α-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphorylcholine, 1-Palmitoyl-2-oleoyl-sn-glycero-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-glycero-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glyceryl-3-phosphorylcholine, 1-Palmitoyl-2-oleoylphosphatidylcholine, 1-Palmitoyl-2-oleyl-3-sn-phosphatidylcholine, 2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, 3,5,8-Trioxa-4-phosphahexacos-17-en-1-aminium, 4-hydroxy-N,N,N-trimethyl-9-oxo-7-[[(1-oxohexadecyl)oxy]methyl]-, inner salt, 4-oxide, [R-(Z)]-, L-1-Palmitoyl-2-oleoylphosphatidylcholine, L-Palmitoyloleoylphosphatidylcholine, L-α-1-Palmitoyl-2-oleoylphosphatidylcholine, POPC, Palmitoyloleoylphosphatidylcholine, β-Oleoyl-γ-palmitoyl-L-α-phosphatidylcholine
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(Z)-octadec-9-enoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCC\C=C/CCCCCCCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1
Formula Weight 759.5778
Chemical Name or Material 1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.