missing translation for 'onlineSavingsMsg'
Learn More

1-Pentadecanol, 99%

Product Code. 15458819
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458819 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458819 Supplier Thermo Scientific Alfa Aesar Supplier No. L03673.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 629-76-5 | C15H32O | 228.42 g/mol

1-Pentadecanol is employed as starting reagent for the asymmetric synthesis of jaspine. It acts as a fatty alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentadecanol is employed as starting reagent for the asymmetric synthesis of jaspine. It acts as a fatty alcohol.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing a
TRUSTED_SUSTAINABILITY

Specifications

CAS 629-76-5
Melting Point 45°C to 46°C
Boiling Point 269°C to 271°C
Molecular Formula C15H32O
MDL Number MFCD00004759
Quantity 5 g
Beilstein 1700695
Synonym 1-pentadecanol, pentadecyl alcohol, pentadecanol, n-pentadecanol, n-1-pentadecanol, neodol 5, unii-333qva4g2q, dsstox_cid_7270, dsstox_rid_78381, dsstox_gsid_27270
Solubility Information Insoluble in water.
InChI Key REIUXOLGHVXAEO-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCO
IUPAC Name pentadecan-1-ol
Molecular Weight (g/mol) 228.42
PubChem CID 12397
ChEBI CHEBI:77468
Formula Weight 228.42
Percent Purity 99%
Chemical Name or Material 1-Pentadecanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.