missing translation for 'onlineSavingsMsg'
Learn More

1-Pentadecanol, TRC

Product Code. 30410998
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410998 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410998 Supplier Toronto Research Chemicals Supplier No. TRCP22044025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Pentadecanol
CAS 629-76-5
Molecular Formula C15 H32 O
Quantity 25 g
Synonym 1-Pentadecanol (7CI','8CI','9CI','ACI), Pentadecanol (6CI), Alfol 15, n-1-Pentadecanol, n-Pentadecanol, NSC 66446, Pentadecyl alcohol
InChI Formula InChI=1S/C15H32O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h16H,2-15H2,1H3
SMILES CCCCCCCCCCCCCCCO
IUPAC Name pentadecan-1-ol
Molecular Weight (g/mol) 228.41
Formula Weight 228.2453
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.