missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanol, 98+%

Product Code. 11414424
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414424 500 mL 500mL
11424424 2500 mL 2500mL
11434424 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414424 Supplier Thermo Scientific Alfa Aesar Supplier No. A13093.AP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71-41-0 | C5H12O | 88.15 g/mol

1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.

Solubility
Miscible with water, alcohol, ether, acetone and most organic solvents.

Notes
Incompatible with strong oxidizing agents, alkali metals, strong acids, halides and aldehydes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Melting Point -79°C
Density 0.812
Boiling Point 136°C to 138°C
Flash Point 49°C (120°F)
Odor Mild
Linear Formula CH3(CH2)4OH
Refractive Index 1.4103
Quantity 500 mL
UN Number UN1105
Beilstein 1730975
Merck Index 14,7118
Solubility Information Miscible with water,alcohol,ether,acetone and most organic solvents.
Formula Weight 88.15
Percent Purity ≥98%
Chemical Name or Material 1-Pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Pentanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 1-Pentanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.