missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanol, 99%, pure

Product Code. 10184170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10184170 25 mL Glass Bottle 25mL
10752581 1 L Glass Bottle 1 L
10215590 2.5 L Glass Bottle 2.5 L
10013313 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10184170 Supplier Thermo Scientific Acros Supplier No. 160600250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71-41-0 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -78.0°C
Density 0.8110g/mL
Boiling Point 137.0°C to 139.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)4OH
Refractive Index 1.4083 to 1.4103
Quantity 25 mL
Beilstein 01, 383
Merck Index 15, 7230
Specific Gravity 0.811
Solubility Information Solubility in water: 22g/L (22°C). Other solubilities: miscible with alcohol, benzene and ether, soluble in most organic solvents
Viscosity 4 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1-Pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Pentanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.