missing translation for 'onlineSavingsMsg'
Learn More

1-Penten-3-ol, TRC

Product Code. 30179017
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179017 1 g 1g
30178969 500 mg 500mg
30179648 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179017 Supplier Toronto Research Chemicals Supplier No. TRCP2275051G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-25-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
Synonym (±)-1-Penten-3-ol, 1-Ethylallyl alcohol, 3-Hydroxy-1-pentene, Ethyl vinyl carbinol, NSC 65446, α-Ethylallyl alcohol
IUPAC Name pent-1-en-3-ol
SMILES CCC(O)C=C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H10O/c1-3-5(6)4-2/h3,5-6H,1,4H2,2H3
Formula Weight 86.0732
Chemical Name or Material 1-Penten-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.