missing translation for 'onlineSavingsMsg'
Learn More

1-Pentene, 97%, AcroSeal™

Product Code. 10309847 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10309847 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10309847 Supplier Thermo Scientific Acros Supplier No. 430731000
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-67-1 | C5H10 | 70.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 109-67-1
CAS Min % 96.0
CAS Max % 100.0
Melting Point -165.0°C
Color Colorless
Density 0.6400g/mL
Boiling Point 30.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C5H10
Linear Formula CH3CH2CH2CH=CH2
Refractive Index 1.3704 to 1.3724
MDL Number MFCD00003567
Quantity 100 mL
Merck Index 15,7234
Specific Gravity 0.64
Synonym 1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether and beznene, soluble in most organic solvents
InChI Key YWAKXRMUMFPDSH-UHFFFAOYSA-N
SMILES CCCC=C
IUPAC Name pent-1-ene
Molecular Weight (g/mol) 70.14
PubChem CID 8004
Viscosity 0.202 mPa.s (20°C)
Formula Weight 70.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Pentene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.