missing translation for 'onlineSavingsMsg'
Learn More

1-Pentene, 97%

Product Code. 11482728
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482728 100 g 100g
11472728 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482728 Supplier Thermo Scientific Alfa Aesar Supplier No. A16723.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 109-67-1 | C5H10 | 70.14 g/mol

1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.

Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, reducing agents, halogens and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-67-1
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00003567
InChI Key YWAKXRMUMFPDSH-UHFFFAOYSA-N
Synonym 1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
PubChem CID 8004
IUPAC Name pent-1-ene
SMILES CCCC=C

Specifications

Melting Point -138°C
Density 0.64
Boiling Point 30°C to 31°C
Flash Point −51°C (−60°F)
Refractive Index 1.3715
Quantity 100 g
UN Number UN1108
Beilstein 1731629
Merck Index 14,7122
Solubility Information Miscible with ethanol,ethyl ether,benzene and organic solvents. Immiscible with water.
Formula Weight 70.14
Percent Purity 97%
Chemical Name or Material 1-Pentene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.