missing translation for 'onlineSavingsMsg'
Learn More

1-Pentyl isocyanate, 97%

Product Code. 11383027
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383027 1 g 1g
11393027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383027 Supplier Thermo Scientific Alfa Aesar Supplier No. L13399.03
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3954-13-0 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3954-13-0
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00144846
InChI Key VRVUKQWNRPNACD-UHFFFAOYSA-N
Synonym pentyl isocyanate, pentylisocyanate, pentane, 1-isocyanato, 1-pentyl isocyanate, n-pentyl isocyanate, 1-isocyanato-pentane, amylisocyanat, n-pentylisocyanate, n-amyl isocyanate, acmc-1clhg
PubChem CID 206036
IUPAC Name 1-isocyanatopentane
SMILES CCCCCN=C=O

Specifications

Density 0.878
Boiling Point 136°C to 137°C
Flash Point 32°C (89°F)
Odor Pungent
Linear Formula CH3(CH2)4NCO
Refractive Index 1.415
Quantity 1 g
UN Number UN3080
Beilstein 1746688
Sensitivity Moisture sensitive
Formula Weight 113.16
Percent Purity 97%
Chemical Name or Material 1-Pentyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.