missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1,4-pentanedione, 96%

Product Code. 11377936
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377936 5 g 5g
11367936 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377936 Supplier Thermo Scientific Alfa Aesar Supplier No. L13151.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 583-05-1 | C11H12O2 | 176.215 g/mol

1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-05-1
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00092889
InChI Key RBLXWIPBPPVLPU-UHFFFAOYSA-N
Synonym 1-phenyl-1,4-pentanedione, 1-phenyl-pentane-1,4-dione, 1,4-pentanedione, 1-phenyl, acmc-20aosh, 1-phenyl-1,4-pentandione, 5-phenyl-2,5-pentanedione, benzil-related compound, 41, 1-phenyl pentane-1,4-dione
PubChem CID 136371
IUPAC Name 1-phenylpentane-1,4-dione
SMILES CC(=O)CCC(=O)C1=CC=CC=C1

Specifications

Boiling Point 158°C to 162°C (12 mmHg)
Refractive Index 1.533
Quantity 5 g
Beilstein 908230
Solubility Information Not miscible in water.
Formula Weight 176.22
Percent Purity 96%
Chemical Name or Material 1-Phenyl-1,4-pentanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.