missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-butyne, 98%

Product Code. 11444150
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444150 5 g 5g
11463734 10 g 10g
11454150 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444150 Supplier Thermo Scientific Alfa Aesar Supplier No. A11332.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 622-76-4 | C10H10 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-76-4
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00039945
InChI Key FFFMSANAQQVUJA-UHFFFAOYSA-N
Synonym 1-phenyl-1-butyne, 1-butynylbenzene, benzene, 1-butynyl, 1-butyne, 1-phenyl, but-1-yn-1-ylbenzene, 1-but-1-ynyl benzene, 1-ethyl-2-phenylacetylene, phenylbutyne, ethylphenylacetylene, phenylethylacetylene
PubChem CID 69328
IUPAC Name but-1-ynylbenzene
SMILES CCC#CC1=CC=CC=C1

Specifications

Density 0.918
Boiling Point 201°C to 203°C
Flash Point 68°C (154°F)
Linear Formula CH3CH2C≡CC6H5
Refractive Index 1.552
Quantity 5 g
Beilstein 1903397
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 1-Phenyl-1-butyne
compliance-icons
Product Identifier
  • 1-Phenyl-1-butyne
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Butynylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.