missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclohexanecarboxylic acid, 95%

Product Code. 10409300 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10409300 50 g Glass bottle 50g
10205990 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10409300 Supplier Thermo Scientific Acros Supplier No. 170750500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1135-67-7 | C13H16O2 | 204.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-67-7
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.27
MDL Number MFCD00037152
InChI Key QXXHHHWXFHPNOS-UHFFFAOYSA-N
Synonym 1-phenylcyclohexanecarboxylic acid, 1-phenyl-1-cyclohexanecarboxylic acid, 1-phenyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-phenyl, acmc-1c6ij, 1-phenylcyclohexanecarboxylicacid, qxxhhhwxfhpnos-uhfffaoysa, 1-phenylcyclohexanecarboxylic acid #, cyclohexanecarboxylicacid, 1-phenyl, 1-phenyl-1-cyclohexane carboxylic acid
PubChem CID 70817
IUPAC Name 1-phenylcyclohexane-1-carboxylic acid
SMILES C1CCC(CC1)(C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 119°C to 124°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula C6H5C6H10CO2H
Quantity 50 g
Formula Weight 204.27
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1-Phenyl-1-cyclohexanecarboxylic acid, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.