missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclopropanecarbonitrile, 97%

Product Code. 10174280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10174280 10 g Glass bottle 10g
10184280 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174280 Supplier Thermo Scientific Acros Supplier No. 170590100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 935-44-4 | C10H9N | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-44-4
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00001270
InChI Key ZHFURHRJUWYDKG-UHFFFAOYSA-N
Synonym 1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
PubChem CID 70288
IUPAC Name 1-phenylcyclopropane-1-carbonitrile
SMILES N#CC1(CC1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 133.0°C to 137.0°C (30.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C3H4CN
Refractive Index 1.5370 to 1.5400
Quantity 10 g
Specific Gravity 1
Formula Weight 143.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-cyclopropanecarbonitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenyl-1-cyclopropanecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.