missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclopropanecarboxylic acid, 97%

Product Code. 10307500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307500 10 g Glass bottle 10g
10367540 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10307500 Supplier Thermo Scientific Acros Supplier No. 170680100
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 6120-95-2 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6120-95-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00001288
InChI Key IWWCCNVRNHTGLV-UHFFFAOYSA-N
Synonym 1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid
PubChem CID 80206
IUPAC Name 1-phenylcyclopropane-1-carboxylic acid
SMILES C1CC1(C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 84°C to 87°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C3H4CO2H
Quantity 10 g
Formula Weight 162.19
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1-Phenyl-1-cyclopropanecarboxylic acid, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.