missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-pentyne, 98%

Product Code. 11485806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485806 5 g 5g
11495806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485806 Supplier Thermo Scientific Alfa Aesar Supplier No. B20857.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4250-81-1 | C11H12 | 144.22 g/mol

It finds it application in the highly regioselective synthesis of polysubstituted naphthalene derivatives through gallium trichloride catalyzed alkyne-aldehyde coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application in the highly regioselective synthesis of polysubstituted naphthalene derivatives through gallium trichloride catalyzed alkyne-aldehyde coupling.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4250-81-1
Molecular Formula C11H12
Molecular Weight (g/mol) 144.22
MDL Number MFCD00039959
InChI Key DEGIOKWPYFOHGH-UHFFFAOYSA-N
Synonym 1-phenyl-1-pentyne, pent-1-yn-1-ylbenzene, 1-pentynylbenzene, benzene, 1-pentynyl, 1-pentynylbenzene #, acmc-1aso7, 1-phenyl-2-propylacetylene
PubChem CID 77923
IUPAC Name pent-1-ynylbenzene
SMILES CCCC#CC1=CC=CC=C1

Specifications

Density 0.906
Boiling Point 213°C to 215°C
Flash Point 82°C (179°F)
Linear Formula CH3CH2CH2C≡CC6H5
Refractive Index 1.5413
Quantity 5 g
Beilstein 1853156
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 1-Phenyl-1-pentyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.