missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-propyne, 99%

Product Code. 10177713 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10177713 25 g Glass bottle 25g
10124190 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10177713 Supplier Thermo Scientific Acros Supplier No. 171320250
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 673-32-5 | C9H8 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-32-5
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009272
InChI Key GHUURDQYRGVEHX-UHFFFAOYSA-N
Synonym 1-phenyl-1-propyne, prop-1-yn-1-ylbenzene, benzene, 1-propynyl, methylphenylacetylene, 2-propynylbenzene, 1-phenylpropyne, 1-phenylpropyne-1, unii-448376bfqc, prop-1-ynyl-benzene, 1-methyl-2-phenylacetylene
PubChem CID 69601
IUPAC Name prop-1-ynylbenzene
SMILES CC#CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.9200g/mL
Boiling Point 185.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C≡CCH3
Refractive Index 1.5630 to 1.5650
Quantity 25 g
Beilstein 05, 514
Specific Gravity 0.92
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-propyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.