missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1H-pyrazole-4-carboxaldehyde, 98%

Product Code. 15430529
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430529 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15430529 Supplier Thermo Scientific Alfa Aesar Supplier No. H63695.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 54605-72-0 | C10H8N2O | 172.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54605-72-0
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.19
MDL Number MFCD02179568
InChI Key PHVRLPFVPVKYOI-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazole-4-carbaldehyde, 1-phenyl-1h-pyrazole-4-carboxaldehyde, 1-phenylpyrazole-4-carboxaldehyde, pubchem13271, acmc-1awxu, phenyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,1-phenyl
PubChem CID 952089
IUPAC Name 1-phenylpyrazole-4-carbaldehyde
SMILES O=CC1=CN(N=C1)C1=CC=CC=C1

Specifications

Melting Point 84°C to 88°C
Color Yellow
Quantity 250 mg
Sensitivity Light and moisture sensitive
Formula Weight 172.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Phenyl-1H-pyrazole-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.