missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10324192
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10324192 250 mg Amber Glass Bottle 250mg
10160782 1 g Amber Glass Bottle 1g
10310494 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10324192 Supplier Thermo Scientific Maybridge Supplier No. CC24501CB
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1133-77-3
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00463580
InChI Key FLYDUXCFCARXHI-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazole-5-carboxylic acid, 2-phenyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-phenyl, 1-phenylpyrazole-5-carboxylic acid, 1-phenyl-1h-pyrazole-5-carboxylicacid, 1h-pyrazole-5-carboxylicacid, 1-phenyl, 5-carboxy-1h-pyrazol-1-yl benzene
PubChem CID 237056
IUPAC Name 2-phenylpyrazole-3-carboxylic acid
SMILES OC(=O)C1=CC=NN1C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 1-Phenyl-1H-pyrazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.