missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1H-pyrazole, 98%

Product Code. 11492417
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492417 50 g 50g
11482417 10 g 10g
11402427 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492417 Supplier Thermo Scientific Alfa Aesar Supplier No. A14270.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1126-00-7 | C9H8N2 | 144.18 g/mol

1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126-00-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00003112
InChI Key WITMXBRCQWOZPX-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazole, n-phenylpyrazole, 1h-pyrazole, 1-phenyl, pyrazole, 1-phenyl, 1-phenylpyrazol, n-phenyl pyrazole, 1-phenyl pyrazole, 1-phenyl-pyrazole, 1-phenyl-1h-pyrazol, l-phenyl-1h-pyrazole
PubChem CID 70769
ChEBI CHEBI:38879
IUPAC Name 1-phenylpyrazole
SMILES C1=CN(N=C1)C1=CC=CC=C1

Specifications

Density 1.09
Boiling Point 141°C to 142°C (30 mmHg)
Flash Point 109°C (228°F)
Refractive Index 1.596
Quantity 50 g
Beilstein 2594
Solubility Information Soluble in water.
Formula Weight 144.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1H-pyrazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.