missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1H-tetrazole-5-thiol, 99%

Product Code. 11400523
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400523 100 g 100g
11490513 25 g 25g
11410523 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400523 Supplier Thermo Scientific Alfa Aesar Supplier No. A12656.22
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 86-93-1 | C7H6N4S | 178.21 g/mol

1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.

Solubility
Soluble in 5% ethanol, water (partly), acetone, chloroform, and methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-93-1
Molecular Formula C7H6N4S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00003129
InChI Key GGZHVNZHFYCSEV-UHFFFAOYSA-N
Synonym 1-phenyl-1h-tetrazole-5-thiol, 1-phenyltetrazole-5-thiol, 1-phenyl-5-mercaptotetrazole, 5-mercapto-1-phenyltetrazole, 1-phenyltetrazoline-5-thione, mercaptophenyltetrazole, phenylmercaptotetrazole, 1-phenyl-5-tetrazolethione, 5-mercapto-1-phenyltetrazol, 1-phenyltetrazole-thiol
PubChem CID 690730
ChEBI CHEBI:79916
SMILES S=C1N=NNN1C1=CC=CC=C1

Specifications

Melting Point ∼145°C (decomposition)
Flash Point 138°C (280°F)
Odor Odorless
Quantity 100 g
UN Number UN1325
Beilstein 139068
Solubility Information Soluble in 5% ethanol,water (partly),acetone,chloroform,and methanol.
IUPAC Name 1-phenyl-2H-tetrazole-5-thione
Formula Weight 178.22
Percent Purity 99%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 1-Phenyl-1H-tetrazole-5-thiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Phenyl-1H-tetrazole-5-thiol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: 1-Phenyl-1H-tetrazole-5-thiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.