missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3,4-dihydro-isoquinoline, TRC

Product Code. 30421668
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421668 500 mg
30415364 250 mg
30416519 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421668 Supplier Toronto Research Chemicals Supplier No. TRCP229440500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52250-50-7
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.27
Synonym 3,4-Dihydro-1-phenylisoquinoline, 1-Phenyl-3,4-dihydroisoquinoline
IUPAC Name 1-phenyl-3,4-dihydroisoquinoline
SMILES C1Cc2ccccc2C(=N1)c3ccccc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H13N/c1-2-7-13(8-3-1)15-14-9-5-4-6-12(14)10-11-16-15/h1-9H,10-11H2
Formula Weight 207.1048
Chemical Name or Material 1-Phenyl-3,4-dihydro-isoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.