missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-pyrazolidinone 97%, ACROS Organics™

Product Code. 10627512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627512 100 g Plastic bottle 100g
10617322 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10627512 Supplier Thermo Scientific Acros Supplier No. 155981000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 92-43-3
Molecular Formula C9H10N2O
Molecular Weight (g/mol) 162.192
MDL Number MFCD00003094
InChI Key CMCWWLVWPDLCRM-UHFFFAOYSA-N
Synonym phenidone, 1-phenyl-3-pyrazolidone, fenidon, 1-phenyl-3-pyrazolidinone, 3-pyrazolidinone, 1-phenyl, 1-phenyl-3-oxopyrazolidine, 2-pyrazolin-3-ol, 1-phenyl, pyrazolidinone, 1-phenyl, 1-phenyl-pyrazolidin-3-one, unii-h0u5612p6k
PubChem CID 7090
IUPAC Name 1-phenylpyrazolidin-3-one
SMILES C1CN(NC1=O)C2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 119°C to 123°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 100 g
Beilstein 24, 2
Merck Index 15, 7419
Formula Weight 162.19
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-3-pyrazolidinone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Phenyl-3-pyrazolidinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.