missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-5-(pyridin-2-yl)-2(1H)-pyridone, TRC

Product Code. 30410800
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410800 1 g -
30428544 2.5 g 2.5g
30428060 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410800 Supplier Toronto Research Chemicals Supplier No. TRCP3367501G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381725-50-4
Molecular Formula C16 H12 N2 O
Molecular Weight (g/mol) 248.279
Synonym 1'-Phenyl-[2,3'-bipyridin]-6'(1'H)-one, 5-(2-Pyridyl)-1-phenyl-1,2-dihydropyridin-2-one
IUPAC Name 1-phenyl-5-pyridin-2-ylpyridin-2-one
SMILES O=C1C=CC(=CN1c2ccccc2)c3ccccn3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H12N2O/c19-16-10-9-13(15-8-4-5-11-17-15)12-18(16)14-6-2-1-3-7-14/h1-12H
Formula Weight 248.095
Chemical Name or Material 1-Phenyl-5-(pyridin-2-yl)-2(1H)-pyridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.