missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyldodecane, 97%

Product Code. 10491163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10491163 50 g Glass bottle 50g
10317900 10 g Glass bottle 10g
10045823 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10491163 Supplier Thermo Scientific Acros Supplier No. 151020500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-01-3 | C18H30 | 246.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-01-3
Molecular Formula C18H30
Molecular Weight (g/mol) 246.43
MDL Number MFCD00008974
InChI Key KWKXNDCHNDYVRT-UHFFFAOYSA-N
Synonym 1-phenyldodecane, benzene, dodecyl, n-dodecylbenzene, laurylbenzene, phenyldodecane, marlican, dodecane, 1-phenyl, detergent alkylate, alkylate p 1, phenyldodecan
PubChem CID 31237
IUPAC Name dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -7°C
Density 0.8500g/mL
Boiling Point 288°C to 300°C
Flash Point 124°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5(CH2)11CH3
Refractive Index 1.4810 to 1.483
Quantity 50 g
Specific Gravity 0.85
Solubility Information Solubility in water: immiscible with water
Formula Weight 246.43
Percent Purity 97%
Chemical Name or Material 1-Phenyldodecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.