missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-1-Phenylethanol-d10, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Phenylethanol-D10 |
| CAS | 219586-41-1 |
| Molecular Formula | C8 D10 O |
| Quantity | 0.5 g |
| Synonym | Benzene-d5-methan-d-ol-d, α-(methyl-d3)- (9 CI), 1-Phenylethanol-d10, α-(Methyl-d3)benzene-d5-methan-d-ol-d, α-(Methyl-d3)benzene-2,3,4,5,6-d5-methan-d-ol-d, O,1,2,2,2-Pentadeuterio-1-(2,3,4,5,6-pentadeuteriophenyl)ethanol, (±)-1-Phenylethanol-d10, (±)-1-Phenylethanol-d10 |
| InChI Formula | InChI=1 S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9 H,1H3/i1D3,2 D,3 D,4 D,5 D,6 D,7 D,9 D |
| SMILES | [2 H]OC([2 H])(c1c([2 H])c([2 H])c([2 H])c([2 H])c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(1,2,2,2-tetradeuterio-1-deuteriooxyethyl)benzene |
| Molecular Weight (g/mol) | 132.226 |
| Formula Weight | 132.1359 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction