missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylethyl mercaptan, 98%

Product Code. 11481240
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481240 1 g 1g
11491240 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481240 Supplier Thermo Scientific Alfa Aesar Supplier No. H26345.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6263-65-6 | C8H10S | 138.228 g/mol

The compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.

Solubility
Insoluble in water. Solubility in ethanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6263-65-6
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00053515
InChI Key QZZBJCFNHPYNKO-UHFFFAOYSA-N
Synonym 1-phenylethyl mercaptan, benzenemethanethiol, .alpha.-methyl, +/--1-phenylethylmercaptan, 1-phenylethane-1-thiol, phenylethanethiol, benzenemethanethiol, alpha-methyl, 1-phenyl ethanethiol, 1-phenyl-ethanethiol, 1-phenethyl mercaptan, 1-phenylethylmercaptan
PubChem CID 141850
IUPAC Name 1-phenylethanethiol
SMILES CC(C1=CC=CC=C1)S

Specifications

Density 1.021
Flash Point 76°C (169°F)
Refractive Index 1.558
Quantity 1 g
UN Number UN2810
Sensitivity Air sensitive
Solubility Information Insoluble in water. Solubility in ethanol.
Formula Weight 138.23
Percent Purity 98%
Chemical Name or Material 1-Phenylethyl mercaptan
compliance-icons
Product Identifier
  • 1-Phenylethyl mercaptan
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Phenylethyl mercaptan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.