missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylnaphthalene, 97%

Product Code. 11445057
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445057 5 g 5g
11455057 25 g 25g
11465057 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445057 Supplier Thermo Scientific Alfa Aesar Supplier No. B21953.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 605-02-7 | C16H12 | 204.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-02-7
Molecular Formula C16H12
Molecular Weight (g/mol) 204.27
MDL Number MFCD00003983
InChI Key IYDMICQAKLQHLA-UHFFFAOYSA-N
Synonym naphthalene, 1-phenyl, 1-phenyl-naphthalene, unii-10nxc4g45q, phenylnaphthalene, phenyl-napthalene, p-4-naphthyl benzene, acmc-1bfcg, 1-phenylnaphthalene, naphthalene, 1-phenyl-8ci 9ci, 10 microg/ml in acetonitrile
PubChem CID 11795
IUPAC Name 1-phenylnaphthalene
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=CC=C1

Specifications

Density 1.088
Boiling Point 320°C
Flash Point >110°C (230°F)
Refractive Index 1.664
Quantity 5 g
Beilstein 1906885
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 1-Phenylnaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.