missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpiperazine, 97%

Product Code. 10657102 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10657102 100 g Glass bottle 100g
10157893 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10657102 Supplier Thermo Scientific Acros Supplier No. 130821000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-54-6 | C10H14N2 | 162.24 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-54-6
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.24
MDL Number MFCD00005957
InChI Key YZTJYBJCZXZGCT-UHFFFAOYSA-N
Synonym phenylpiperazine, n-phenylpiperazine, piperazine, 1-phenyl, 1-phenyl-piperazine, n-phenyldiethylenediamine, 1-fenylpiperazin czech, unii-j9225cbi7d, n-phenyl piperazine, 1-phenyl piperazine, ccris 4334
PubChem CID 7096
IUPAC Name 1-phenylpiperazine
SMILES C1CN(CCN1)C2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 18.8°C
Color Colorless to Yellow
Density 1.0620g/mL
Boiling Point 286.0°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. (Aniline) (GC)
Packaging Glass bottle
Refractive Index 1.5865 to 1.5885
Quantity 100 g
UN Number 2927
Specific Gravity 1.062
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 162.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Phenylpiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.