missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpiperazine, 98+%

Product Code. 11494344
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494344 100 g 100g
11484344 25 g 25g
11404354 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494344 Supplier Thermo Scientific Alfa Aesar Supplier No. A12610.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-54-6 | C10H14N2 | 162.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-54-6
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD00005957
InChI Key YZTJYBJCZXZGCT-UHFFFAOYSA-N
Synonym phenylpiperazine, n-phenylpiperazine, piperazine, 1-phenyl, 1-phenyl-piperazine, n-phenyldiethylenediamine, 1-fenylpiperazin czech, unii-j9225cbi7d, n-phenyl piperazine, 1-phenyl piperazine, ccris 4334
PubChem CID 7096
IUPAC Name 1-phenylpiperazine
SMILES C1CN(CCN1)C2=CC=CC=C2

Specifications

Melting Point 17°C to 19°C
Density 1.064
Boiling Point 285°C to 286°C
Flash Point 141°C (286°F)
Refractive Index 1.588
Quantity 100 g
UN Number UN2927
Beilstein 132157
Formula Weight 162.24
Percent Purity ≥98%
Chemical Name or Material 1-Phenylpiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.