missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpiperazine hydrochloride, 99%

Product Code. 11488836
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488836 25 g 25g
11498836 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488836 Supplier Thermo Scientific Alfa Aesar Supplier No. A16995.14
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2210-93-7 | C10H15ClN2 | 198.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2210-93-7
Molecular Formula C10H15ClN2
Molecular Weight (g/mol) 198.694
MDL Number MFCD00012758
InChI Key NQNZNONJZASOKL-UHFFFAOYSA-N
Synonym 1-phenylpiperazine hydrochloride, 1-phenylpiperazinium chloride, piperazine, 1-phenyl-, hydrochloride, phenylpiperazine hydrochloride, 1-phenyl piperazine hydrochloride, piperazine, 1-phenyl-, monohydrochloride, phenylpiperazine, chloride, 1-phenylpiperazine hcl, c10h14n2.hcl, 1-phenylpiperazinehydrochloride
PubChem CID 75164
IUPAC Name 1-phenylpiperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=CC=C2.Cl

Specifications

Melting Point 247°C to 250°C
Odor Odorless
Quantity 25 g
UN Number UN2811
Formula Weight 198.7
Percent Purity 99%
Chemical Name or Material 1-Phenylpiperazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.