missing translation for 'onlineSavingsMsg'
Learn More

1-Piperidinepropionic acid, 96%, Thermo Scientific Chemicals

Product Code. 11986391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11986391 100 g 100g
11976391 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986391 Supplier Thermo Scientific Acros Supplier No. 440051000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26371-07-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00006515
InChI Key LPDGWMLCUHULJF-UHFFFAOYSA-N
Synonym 1-piperidinepropionic acid, 1-piperidinepropanoic acid, 3-piperidin-1-yl propanoic acid, unii-6l69a7vpho, piperidine-1-propionic acid, 3-1-piperidinyl propanoic acid, 3-piperidinopropanoic acid, 6l69a7vpho, piperidinepropionic acid, 3-1-piperidinyl propionic acid
PubChem CID 117782
SMILES OC(=O)CCN1CCCCC1

Specifications

Melting Point 86.0°C to 88.5°C
CAS Min % 95.0
CAS Max % 100.0
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
IUPAC Name 3-(piperidin-1-yl)propanoic acid
Quantity 100 g
Formula Weight 157.21
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1-Piperidinepropionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1-Piperidinepropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.