missing translation for 'onlineSavingsMsg'
Learn More

1-Propanesulfonic acid, 95%

Product Code. 10329343 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10329343 5 g Glass bottle 5g
10158180 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10329343 Supplier Thermo Scientific Acros Supplier No. 418340050
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5284-66-2 | C3H8O3S | 124.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5284-66-2
Molecular Formula C3H8O3S
Molecular Weight (g/mol) 124.16
MDL Number MFCD00041889
InChI Key KCXFHTAICRTXLI-UHFFFAOYSA-N
Synonym 1-propanesulfonic acid, propanesulfonic acid, propylsulfonic acid, propanesulfonate, propanesulphonic acid, ammonium propanesulfonate, propane-1-sulphonic acid, ncistruc1_001874, ncistruc2_000257, 1-propanesulfonic acid, technical t
PubChem CID 78938
IUPAC Name propane-1-sulfonic acid
SMILES CCCS(=O)(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 8.0°C
Color Brown
Density 1.2500g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3CH2CH2SO3H
Refractive Index 1.4380 to 1.4410
Quantity 5 g
Specific Gravity 1.25
Formula Weight 124.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Propanesulfonic acid, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.