missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol ≥ 99.0%, BAKER ANALYZED™, J.T. Baker™

Product Code. 10280901 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10280901 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10280901 Supplier JT Baker Supplier No. 8066.1000
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratory Reagent

Specifications

Melting Point -127°C
CAS 71-23-8
Color Max. 10 (APHA)
Density 0.8053 g/cm³ (20°C)
Quantity 1 L
Flash Point 15°C
Packaging Glass Bottle
Residue after Evaporation max. 0.005%
Assay Percent Range min. 99%
Molecular Formula C3H8O
MDL Number MFCD00002941
Water max. 0.1%
Boiling Point 97°C (1013 hPa)
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
SMILES CCCO
IUPAC Name propan-1-ol
Molecular Weight (g/mol) 60.10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.