missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, 99.5%, Extra Dry, AcroSeal™

Product Code. 10592944 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10592944 1 L 1 L
10003493 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10592944 Supplier Thermo Scientific Acros Supplier No. 396940010
  • ONLINE EXCLUSIVE PRICE
    374.00 €
    176.00 € / 1 L
    Save 198.00 €
    52% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71-23-8 | C3H8O | 60.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Spezifikation

Melting Point -127.0°C
CAS Min % 99.4
CAS Max % 100.0
Density 0.8000g/mL
Quantity 1 L
Flash Point 15°C
Infrared Spectrum Authentic
Physical Form Liquid
Refractive Index 1.3840 to 1.3860
Residue after Evaporation 0.001 % max.
Assay Percent Range 99.5%
Specific Gravity 0.8
Linear Formula CH3CH2CH2OH
Viscosity 2.2 mPa.s (20°C)
Water 0.005% Max. (Coulometric)
Merck Index 15, 7955
Boiling Point 97.0°C
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
Formula Weight 60.1
Percent Purity 99.5%
Grade Extra Dry
Chemical Name or Material 1-Propanol, Extra Dry
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • 1-Propanol
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H318-Causes serious eye damage.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.