missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15598524 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
Unit Size:
4 L
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15598524 500 mL Narrow Mouth Amber Glass Bottle Each
15508534 4 L Narrow Mouth PVC-coated Amber Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15598524 Supplier Avantor J.T.Baker Supplier No. 908601
  • ONLINE EXCLUSIVE PRICE
    91.70 €
    72.43 € / Pack of 1
    Save 19.27 €
    21% Off
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Quantity 500 mL
Flash Point 23.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Formula Weight 60.1
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material 1-Propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.