missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haen

Product Code. 15674640
Change view
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2 L
2.5 L
Pack of 4
Pack of 6
Product Code. Quantity Packaging unitSize
15674640 2.5 L Glass Bottle 2.5 L
15654640 100 mL Glass Bottle 100mL
15664640 1 L Glass Bottle 1 L
15684640 2 L Glass Bottle 2 L
15604650 6 x 1 L Glass Bottle Pack of 6
15694640 4 x 2.5 L Glass Bottle Pack of 4
6 options
This item is not returnable. View return policy

Product Code. 15674640

Brand: Honeywell-Chromasolv 348712.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°C (literature)
CAS Max % 100.0000%
Density 0.804 g/mL at 25 °C(lit.)
Boiling Point 96°C to 98°C
Quantity 2.5 L
Flash Point 21°C
Packaging Glass Bottle
Linear Formula CH3CH2CH2OH
UN Number UN1274
Beilstein 1098242
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
Formula Weight 60.1
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material 1-Propanol, CHROMASOLV™, for HPLC, ≥99.9%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.