missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, HPLC Grade, 99% min

Product Code. 11398207
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398207 4 x 1 L 4 L
11378207 1 L 1 L
11388207 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398207 Supplier Thermo Scientific Alfa Aesar Supplier No. 022932.M1
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71-23-8 | C3H8O | 60.10 g/mol

1-Propanol is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point °C to 127°C
Density 0.804
Boiling Point 97°C
Flash Point 15°C (59°F)
Assay Percent Range 99% min.
Odor Alcohol-like
Refractive Index 1.385
Quantity 4 x 1 L
Beilstein 1098242
Solubility Information Miscible with water,alcohols,ketones,aldehydes,ethers,glycols,aromatic and aliphatic hydrocarbons.
Formula Weight 60.1
Vapor Pressure 13.3 hPa (10mm Hg) at 20°C
Grade HPLC
Physical Form Liquid
Chemical Name or Material 1-Propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.