missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, specified according to the requirements of Ph.Eur.

Product Code. 12055380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12055380 2.5 L Glass bottle 2.5 L
12045380 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12055380 Supplier Thermo Scientific Acros Supplier No. 446270025
  • ONLINE EXCLUSIVE PRICE
    146.00 €
    141.00 € / 2.5 L
    Save 5.00 €
    3% Off
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 71-23-8 | C3H8O | 60.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Acidity passes test
Absorbance 0.01 max. at 310nm, 0.02 max. at 290nm, 0.03 max. at 270nm, 0.1 max. at 250nm, 0.3 max. at 230nm
Melting Point -127.0°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 97.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3CH2CH2OH
Refractive Index 1.3840 to 1.3860
Quantity 2.5 L
Merck Index 15,7955
Specific Gravity 0.8
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
Viscosity 2.2 mPa.s (20°C)
Formula Weight 60.1
Percent Purity ≥99.7% (GC)
Related Substances 0.3 (Total impurities) max
Physical Form Liquid
Chemical Name or Material 1-Propanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: n-Propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.