missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, for spectroscopy ACS

Product Code. 10686250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10686250 1 L Glass bottle 1 L
10646630 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10686250 Supplier Thermo Scientific Acros Supplier No. 434360010
  • ONLINE EXCLUSIVE PRICE
    63.80 €
    61.90 € / 1 L
    Save 1.90 €
    2% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71-23-8 | C3H8O | 60.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127.0°C
Density 0.8000g/mL
Boiling Point 97.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3CH2CH2OH
Refractive Index 1.3840 to 1.3860
Quantity 1 L
Residue after Evaporation 0.001% max.
Merck Index 15,7955
Specific Gravity 0.8
Solubility Information Clear colorless
Viscosity 2.2 mPa.s (20°C)
Formula Weight 60.1
Percent Purity ≥99.5%
Grade Spectroscopy ACS
Titratable Acid 0.0004 meq/g
Physical Form Liquid
Chemical Name or Material 1-Propanol, For Spectroscopy
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.